Novus Biologicals
Manufacturer Code:NBP154378
Catalog # NBP154378
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to ACADVL(acyl-Coenzyme A dehydrogenase very long chain) The peptide sequence was selected from the N terminal of ACADVL (NP_001029031). Peptide sequence RPYAGGAAQESKSFAVGMFKGQLTTDQVFPYPSVLNEEQTQFLKELVEPV. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAD6 acyl-CoA dehydrogenase very long chain acyl-Coenzyme A dehydrogenase very long chain EC 1.3.99 EC 1.3.99.- LCACD very long-chain specific acyl-CoA dehydrogenase mitochondrial VLCAD; acyl-Coenzyme A dehydrogenase, very long chain; Very long-chain specific acyl-CoA dehydrogenase, mitochondrial; VLCAD
Gene Aliases: ACAD6; ACADVL; LCACD; VLCAD
UniProt ID: (Human) P49748
Entrez Gene ID: (Human) 37
Molecular Function:
dehydrogenase
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.