Novus Biologicals
Manufacturer Code:NBP152016
Catalog # NBP152016
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence DKSDSHPSDALTRK-C corresponding to N-Terminus (near) according to NP_000009.1 NP_001029031.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAD6 acyl-CoA dehydrogenase very long chain acyl-Coenzyme A dehydrogenase very long chain EC 1.3.99 EC 1.3.99.- LCACD very long-chain specific acyl-CoA dehydrogenase mitochondrial VLCAD; acyl-Coenzyme A dehydrogenase, very long chain; Very long-chain specific acyl-CoA dehydrogenase, mitochondrial; VLCAD
Gene Aliases: ACAD6; ACADVL; LCACD; VLCAD
UniProt ID: (Human) P49748
Entrez Gene ID: (Human) 37
Molecular Function: dehydrogenase oxidase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.