Novus Biologicals
Manufacturer Code:NBP270061UV
Catalog # NBP270061UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D2 |
Immunogen | Recombinant protein fragment corresponding to amino acids 34-260 of human ACADS (NP_000008) produced in E.coli. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ACAD3 acyl-CoA dehydrogenase C-2 to C-3 short chain acyl-Coenzyme A dehydrogenase C-2 to C-3 short chain Butyryl-CoA dehydrogenase EC 1.3.99 EC 1.3.99.2 mitochondrial short-chain specific acyl-CoA dehydrogenase SCADshort-chain acyl-CoA dehydrogenase short-chain specific acyl-CoA dehydrogenase mitochondrial unsaturated acyl-CoA reductase; acyl-Coenzyme A dehydrogenase, C-2 to C-3 short chain; Butyryl-CoA dehydrogenase; mitochondrial short-chain specific acyl-CoA dehydrogenase; SCAD; Short-chain specific acyl-CoA dehydrogenase, mitochondrial; unsaturated acyl-CoA reductase
Gene Aliases: ACAD3; ACADS; SCAD
UniProt ID: (Human) P16219
Entrez Gene ID: (Human) 35
Molecular Function:
dehydrogenase
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.