OriGene
Manufacturer Code:TA811764
Catalog # 50-168-4237
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI10A5 |
Immunogen | Full length human recombit protein of human ACADM produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ACAD1 MCAD MCADH; acyl-CoA dehydrogenase C-4 to C-12 straight chain; acyl-Coenzyme A dehydrogenase, C-4 to C-12 straight chain; MCAD; MCADH; Medium chain acyl-CoA dehydrogenase; Medium-chain specific acyl-CoA dehydrogenase, mitochondrial; testicular tissue protein Li 7
Gene Aliases: ACAD1; ACADM; MCAD; MCADH
UniProt ID: (Human) P11310
Entrez Gene ID: (Human) 34
Molecular Function:
dehydrogenase
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.