Novus Biologicals
Manufacturer Code:NBP154729
Catalog # NBP154729
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to ACADM(acyl-Coenzyme A dehydrogenase C-4 to C-12 straight chain) The peptide sequence was selected from the N terminal of ACADM. Peptide sequence AAGFGRCCRVLRSISRFHWRSQHTKANRQREPGLGFSFEFTEQQKEFQAT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAD1 acyl-CoA dehydrogenase C-4 to C-12 straight chain acyl-Coenzyme A dehydrogenase C-4 to C-12 straight chain EC 1.3.99 EC 1.3.99.3 FLJ18227 FLJ99884 MCADFLJ93013 MCADH medium-chain specific acyl-CoA dehydrogenase mitochondrial; acyl-CoA dehydrogenase C-4 to C-12 straight chain; acyl-Coenzyme A dehydrogenase, C-4 to C-12 straight chain; MCAD; MCADH; Medium chain acyl-CoA dehydrogenase; Medium-chain specific acyl-CoA dehydrogenase, mitochondrial; testicular tissue protein Li 7
Gene Aliases: ACAD1; ACADM; MCAD; MCADH
UniProt ID: (Human) P11310
Entrez Gene ID: (Human) 34
Molecular Function:
dehydrogenase
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.