Invitrogen
Manufacturer Code:PA540792
Catalog # PIPA540792
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ACADL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetyl-Coenzyme A dehydrogenase, long-chain; Acyl Coenzyme A dehydrogenase, long chain; Acyl-CoA dehydrogenase long chain ACAD4 LCAD; acyl-Coenzyme A dehydrogenase, long chain; acyl-Coenzyme A dehydrogenase, long-chain; LCAD; LCAD long chain acyl-CoA dehydrogenase; long-chain acyl-CoA dehydrogenase; Long-chain specific acyl-CoA dehydrogenase, mitochondrial
Gene Aliases: AA960361; ACAD4; ACADL; ACOADA; AU018452; C79855; LCAD; zgc:55656
UniProt ID: (Pig) P79274, (Human) P28330, (Mouse) P51174, (Rat) P15650
Entrez Gene ID: (Rabbit) 100358521, (Bovine) 614508, (Pig) 396931, (Human) 33, (Mouse) 11363, (Rat) 25287, (Horse) 100052267, (Zebrafish) 394156
Molecular Function:
dehydrogenase
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.