Invitrogen
Manufacturer Code:PA527199
Catalog # PIPA527199
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 134 of Human ACADL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAD4 LCAD acyl-Coenzyme A dehydrogenase long chain long-chain specific acyl-CoA dehydrogenase mitochondrial; acyl-Coenzyme A dehydrogenase, long chain; LCAD; Long-chain specific acyl-CoA dehydrogenase, mitochondrial
Gene Aliases: ACAD4; ACADL; LCAD
UniProt ID: (Human) P28330
Entrez Gene ID: (Human) 33
Molecular Function: dehydrogenase oxidase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.