Invitrogen
Manufacturer Code:PA5109332
Catalog # PIPA5109332
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein within N-terminal human ACADL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA960361 ACAD4 ACADL ACOADA AU018452 BOS_2023 C79855 LCAD Long-chain specific acyl-CoA dehydrogenase mitochondrial zgc:55656; acyl-Coenzyme A dehydrogenase, long chain; LCAD; Long-chain specific acyl-CoA dehydrogenase, mitochondrial
Gene Aliases: ACAD4; ACADL; LCAD
UniProt ID: (Human) P28330
Entrez Gene ID: (Human) 33
Molecular Function: dehydrogenase oxidase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.