Invitrogen
Manufacturer Code:PA5103559
Catalog # PIPA5103559
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ACAD9 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2600017P15Rik 4732402K02 ACAD9 ACAD-9 Acyl-CoA dehydrogenase family member 9 acyl-CoA dehydrogenase family member 9 mitochondrial acyl-CoA dehydrogenase family member 9 acyl-Coenzyme A dehydrogenase family member 9 C630012L17Rik Complex I assembly factor ACAD9 mitochondrial NPD002 Nyggf2 very-long-chain acyl-CoA dehydrogenase VLCAD very-long-chain acyl-CoA dehydrogenase VLCAD homolog; ACAD-9; Acyl-CoA dehydrogenase family member 9; acyl-CoA dehydrogenase family member 9, mitochondrial; acyl-CoA dehydrogenase family, member 9; acyl-Coenzyme A dehydrogenase family, member 9; Complex I assembly factor ACAD9, mitochondrial; very-long-chain acyl-CoA dehydrogenase VLCAD; very-long-chain acyl-CoA dehydrogenase VLCAD homolog
Gene Aliases: 2600017P15Rik; 4732402K02; ACAD9; C630012L17Rik; NPD002; Nyggf2
UniProt ID: (Human) Q9H845, (Rat) B1WC61, (Mouse) Q8JZN5
Entrez Gene ID: (Human) 28976, (Rat) 294973, (Mouse) 229211
Molecular Function:
dehydrogenase
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.