Invitrogen
Manufacturer Code:PA521491
Catalog # PIPA521491
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 146 and 415 of ACAD8 (Uniprot IDNo.Q9UKU7) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAD-8; ACAD-8 ARC42 activator-recruited cofactor 42 kDa component acyl-Coenzyme A dehydrogenase family member 8 isobutyryl-CoA dehydrogenase mitochondrial; Activator-recruited cofactor 42 kDa component; Acyl-CoA dehydrogenase family member 8; acyl-CoA dehydrogenase family, member 8; acyl-Coenzyme A dehydrogenase family, member 8; ARC42; IBDH; Isobutyryl-CoA dehydrogenase, mitochondrial
Gene Aliases: ACAD-8; ACAD8; ARC42; IBD
UniProt ID: (Human) Q9UKU7
Entrez Gene ID: (Human) 27034
Molecular Function: dehydrogenase oxidase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.