Novus Biologicals
Manufacturer Code:NBP29230420UL
Catalog # NB026500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human ACAD11 (NP_115545.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAD-11; ACAD-11 acyl-CoA dehydrogenase family member 11 acyl-CoA dehydrogenase family member 11 acyl-Coenzyme A dehydrogenase family member 11 EC 1.3.99.- FLJ12592 MGC150619; Acyl-CoA dehydrogenase family member 11; acyl-CoA dehydrogenase family, member 11; acyl-Coenzyme A dehydrogenase family, member 11
Gene Aliases: ACAD-11; ACAD11
UniProt ID: (Human) Q709F0
Entrez Gene ID: (Human) 84129
Molecular Function: dehydrogenase oxidase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.