Proteintech
Manufacturer Code:171611AP150UL
Catalog # 50-173-1409
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ACAD10 Fusion Protein Ag10510 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAD 10 ACAD10; ACAD-10; Acyl-CoA dehydrogenase family member 10; acyl-CoA dehydrogenase family, member 10; acyl-Coenzyme A dehydrogenase family, member 10
Gene Aliases: 2410021P16Rik; ACAD10; RGD1310159
UniProt ID: (Human) Q6JQN1, (Mouse) Q8K370
Entrez Gene ID: (Human) 80724, (Mouse) 71985, (Rat) 304500
Molecular Function:
dehydrogenase
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.