Invitrogen
Manufacturer Code:PA5114376
Catalog # PIPA5114376
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence of human Acetyl Coenzyme A Carboxylase/ACACB. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACACB ACC2 Accb ACC-beta Acetyl-CoA carboxylase 2 acetyl-CoA carboxylase beta acetyl-Coenzyme A carboxylase beta AI597064 AW743042 Biotin carboxylase HACC275; ACC-beta; Acetyl-CoA carboxylase 2; acetyl-Coenzyme A carboxylase 2; acetyl-Coenzyme A carboxylase beta
Gene Aliases: ACACB; ACC2; ACCB; AI597064; AW743042; HACC275
UniProt ID: (Human) O00763, (Mouse) E9Q4Z2
Entrez Gene ID: (Human) 32, (Rat) 116719, (Mouse) 100705
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.