Novus Biologicals
Manufacturer Code:NBP184974
Catalog # NBP184974
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:LLRGVFVVAAKRTPFGAYGGLLKDFTATDLSEFAAKAALSAGKVSPETVDSVIMGNVLQSSSDAIYLARHVGLRVGIPKETPALTINRLCGSGFQSIVNGCQEICVKEAEVVLCGGTESMSQAPYCVRNVRFGTKLGSDIKLEDSLW |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-ketoacyl-CoA thiolase mitochondrial Acetyl-CoA acyltransferase acetyl-CoA acyltransferase 2 acetyl-Coenzyme A acyltransferase 2 beta ketothiolase beta-ketothiolase DSAEC EC 2.3.1 EC 2.3.1.16 FLJ35992 FLJ95265 Mitochondrial 3-oxoacyl-CoA thiolase mitochondrial 3-oxoacyl-Coenzyme A thiolase T1; 3-ketoacyl-CoA thiolase, mitochondrial; Acetyl-CoA acetyltransferase; Acetyl-CoA acyltransferase; acetyl-Coenzyme A acyltransferase 2; Acyl-CoA hydrolase, mitochondrial; beta ketothiolase; Beta-ketothiolase; Mitochondrial 3-oxoacyl-CoA thiolase; mitochondrial 3-oxoacyl-Coenzyme A thiolase; T1
Gene Aliases: ACAA2; DSAEC
UniProt ID: (Human) P42765
Entrez Gene ID: (Human) 10449
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.