Invitrogen
Manufacturer Code:MA534995
Catalog # PIMA534995
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL2663 |
Immunogen | Recombinant Protein corresonding to Human ACAA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 3-ketoacyl-CoA thiolase A peroxisomal 3-ketoacyl-CoA thiolase B peroxisomal 3-ketoacyl-CoA thiolase peroxisomal ACAA Acaa1 Acaa1a Acaa1b Acetyl-CoA acyltransferase acetyl-CoA acyltransferase 1 acetyl-CoA acyltransferase 1A acetyl-CoA acyltransferase 3-oxo acyl-CoA thiolase A 1 peroxisomal Acetyl-CoA acyltransferase 3-oxo acyl-CoA thiolase A peroxisomal acetyl-CoA acyltransferase A Acetyl-CoA acyltransferase B acetyl-CoA acyltransferase 3-oxo acyl-CoA thiolase A acetyl-Coenzyme A acyltransferase 1 acetyl-Coenzyme A acyltransferase 1 (peroxisomal 3-oxoacyl-Coenzyme A thiolase) acetyl-Coenzyme A acyltransferase 1A acetyl-Coenzyme A acyltransferase 1B beta-ketothiolase Beta-ketothiolase A beta-ketothiolase B D9Ertd25e peroxisomal 3-ketoacyl-CoA thiolase peroxisomal 3-ketoacyl-CoA thiolase B peroxisomal 3-oxoacyl-CoA thiolase Peroxisomal 3-oxoacyl-CoA thiolase A peroxisomal 3-oxoacyl-CoA thiolase B peroxisomal 3-oxoacyl-Coenzyme A thiolase Pktaa PTHIO PTL RGD1562373 testicular tissue protein Li 197 THIO thiolase A thiolase B; 3-ketoacyl-CoA thiolase, peroxisomal; Acetyl-CoA acyltransferase; Acetyl-CoA C-myristoyltransferase; acetyl-Coenzyme A acyltransferase 1; Beta-ketothiolase; Peroxisomal 3-oxoacyl-CoA thiolase; peroxisomal 3-oxoacyl-Coenzyme A thiolase; testicular tissue protein Li 197
Gene Aliases: ACAA; ACAA1; PTHIO; THIO
UniProt ID: (Human) P09110
Entrez Gene ID: (Human) 30
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.