Invitrogen
Manufacturer Code:PA5114806
Catalog # PIPA5114806
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ACAA1 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-ketoacyl-CoA thiolase A peroxisomal 3-ketoacyl-CoA thiolase B peroxisomal 3-ketoacyl-CoA thiolase peroxisomal ACAA Acaa1 Acaa1a Acaa1b Acetyl-CoA acyltransferase acetyl-CoA acyltransferase 1 acetyl-CoA acyltransferase 1A acetyl-CoA acyltransferase 3-oxo acyl-CoA thiolase A 1 peroxisomal Acetyl-CoA acyltransferase 3-oxo acyl-CoA thiolase A peroxisomal acetyl-CoA acyltransferase A Acetyl-CoA acyltransferase B acetyl-CoA acyltransferase 3-oxo acyl-CoA thiolase A acetyl-Coenzyme A acyltransferase 1 acetyl-Coenzyme A acyltransferase 1 (peroxisomal 3-oxoacyl-Coenzyme A thiolase) acetyl-Coenzyme A acyltransferase 1A acetyl-Coenzyme A acyltransferase 1B beta-ketothiolase Beta-ketothiolase A beta-ketothiolase B D9Ertd25e peroxisomal 3-ketoacyl-CoA thiolase peroxisomal 3-ketoacyl-CoA thiolase B peroxisomal 3-oxoacyl-CoA thiolase Peroxisomal 3-oxoacyl-CoA thiolase A peroxisomal 3-oxoacyl-CoA thiolase B peroxisomal 3-oxoacyl-Coenzyme A thiolase Pktaa PTHIO PTL RGD1562373 testicular tissue protein Li 197 THIO thiolase A thiolase B; 3-ketoacyl-CoA thiolase A, peroxisomal; 3-ketoacyl-CoA thiolase B, peroxisomal; 3-ketoacyl-CoA thiolase, peroxisomal; Acetyl-CoA acyltransferase; acetyl-CoA acyltransferase 1A; acetyl-CoA acyltransferase 3-oxo acyl-CoA thiolase A 1 peroxisomal; Acetyl-CoA acyltransferase 3-oxo acyl-CoA thiolase A peroxisomal; Acetyl-CoA acyltransferase A; Acetyl-CoA acyltransferase B; acetyl-CoA acyltransferase, 3-oxo acyl-CoA thiolase A; Acetyl-CoA C-myristoyltransferase; acetyl-Coenzyme A acyltransferase 1; acetyl-Coenzyme A acyltransferase 1 (peroxisomal 3-oxoacyl-Coenzyme A thiolase); acetyl-Coenzyme A acyltransferase 1A; Beta-ketothiolase; Beta-ketothiolase A; Beta-ketothiolase B; peroxisomal 3-ketoacyl-CoA thiolase; peroxisomal 3-ketoacyl-CoA thiolase B; Peroxisomal 3-oxoacyl-CoA thiolase; Peroxisomal 3-oxoacyl-CoA thiolase A; Peroxisomal 3-oxoacyl-CoA thiolase B; peroxisomal 3-oxoacyl-Coenzyme A thiolase; testicular tissue protein Li 197; Thiolase A; thiolase B
Gene Aliases: ACAA; ACAA1; Acaa1a; Acaa1b; D9Ertd25e; Pktaa; PTHIO; PTL; RGD1562373; THIO
UniProt ID: (Human) P09110, (Mouse) Q921H8, (Rat) P21775, (Rat) P07871, (Mouse) Q8VCH0
Entrez Gene ID: (Human) 30, (Mouse) 113868, (Rat) 24157, (Rat) 501072, (Mouse) 235674
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.