Invitrogen
Manufacturer Code:PA571719
Catalog # PIPA571719
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 681-710 amino acids from the C-terminal region of human ABTB2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABTB2A BTBD22 ankyrin repeat and BTB/POZ domain-containing protein 2 ankyrin repeat and BTB (POZ) domain containing 2; ankyrin repeat and BTB (POZ) domain containing 2; Ankyrin repeat and BTB/POZ domain-containing protein 2
Gene Aliases: ABTB2; ABTB2A; BTBD22
UniProt ID: (Human) Q8N961
Entrez Gene ID: (Human) 25841
Molecular Function: actin family cytoskeletal protein cytoskeletal protein growth factor hydrolase non-motor actin binding protein protease serine protease signaling molecule transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.