Invitrogen
Manufacturer Code:PA540569
Catalog # PIPA540569
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ABT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activator of basal transcription 1; Activator of basal transcription 1 hABT1; Basal transcriptional activator; hABT1; TATA-binding protein-binding protein
Gene Aliases: ABT1; hABT1; im:7139827; zgc:109995
UniProt ID: (Human) Q9ULW3, (Bovine) Q148M8, (Rat) Q4KLM5, (Mouse) Q9QYL7
Entrez Gene ID: (Rabbit) 100341869, (Dog) 100856389, (Horse) 100053564, (Human) 29777, (Bovine) 539270, (Rat) 306960, (Guinea pig) 100713553, (Mouse) 30946, (Zebrafish) 553568
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.