Invitrogen
Manufacturer Code:PA5112348
Catalog # PIPA5112348
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Activator of basal transcription 1 protein (197-270AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Abt1 Activator of basal transcription 1 Basal transcriptional activator BOS_21887 hABT1 hypothetical protein LOC553568 im:7139827 LOC100341869 TATA-binding protein-binding protein zgc:109995; Activator of basal transcription 1; Basal transcriptional activator; hABT1; TATA-binding protein-binding protein
Gene Aliases: ABT1; hABT1
UniProt ID: (Human) Q9ULW3, (Mouse) Q9QYL7
Entrez Gene ID: (Human) 29777, (Mouse) 30946
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.