Abnova
Manufacturer Code:H00029777K
Catalog # 89-210-845
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ABT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ABT1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Activator of basal transcription 1; Basal transcriptional activator; hABT1; TATA-binding protein-binding protein
Gene Aliases: ABT1; hABT1
UniProt ID: (Human) Q9ULW3
Entrez Gene ID: (Human) 29777
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.