Novus Biologicals
Manufacturer Code:NBP250485AF647
Catalog # NBP250485AH
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | VA4-20B |
Immunogen | Partially purified lactase blood group A |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A transferase; A transferase A3GALNT A3GALT1 ABO blood group (transferase A alpha 1-3-N-acetylgalactosaminyltransferasetransferase B alpha 1-3-galactosyltransferase) ABO glycosyltransferase B transferase B(A) alpha-13-galactosyltransferase EC 2.4.1.37 EC 2.4.1.40 Fucosylglycoprotein 3-alpha-galactosyltransferase Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase Glycoprotein-fucosylgalactoside alpha-galactosyltransferase Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase GTB Histo-blood group A transferase histo-blood group A2 transferase histo-blood group ABO system transferase Histo-blood group B transferase NAGAT; A1-specific alpha 1-3-N-acetylgalactosaminyltransferase; ABO A3 transferase; ABO glycosyltransferase; ABO weak transfer; B transferase; B(A) alpha-1,3-galactosyltransferase; fucosylglycoprotein; Fucosylglycoprotein 3-alpha-galactosyltransferase; Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase; Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase soluble form; Glycoprotein-fucosylgalactoside alpha-galactosyltransferase; Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase; glycosyltransferase A; glycosyltransferase B; Histo-blood group A transferase; histo-blood group A2 transferase; Histo-blood group ABO system transferase; Histo-blood group B transferase; NAGAT; Transferase; transferase A
Gene Aliases: A3GALNT; A3GALT1; ABO; GTB; NAGAT
UniProt ID: (Human) P16442
Entrez Gene ID: (Human) 28
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.