Abnova
Manufacturer Code:PAB28724
Catalog # 89-130-241
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human ABLIM3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant ABLIM3.
For Research Use Only
Protein Aliases: abLIM-3; actin binding LIM protein family, member 3; Actin-binding LIM protein 3; Actin-binding LIM protein family member 3; HMFN1661
Gene Aliases: ABLIM3; HMFN1661; KIAA0843
UniProt ID: (Human) O94929
Entrez Gene ID: (Human) 22885
Molecular Function: actin family cytoskeletal protein cytoskeletal protein structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.