Proteintech
Manufacturer Code:279811AP150UL
Catalog # 50-173-5799
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ABLIM3 Fusion Protein Ag27578 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: abLIM 3 ABLIM3 Actin binding LIM protein 3 HMFN1661 KIAA0843; abLIM-3; actin binding LIM protein family, member 3; Actin-binding LIM protein 3; Actin-binding LIM protein family member 3
Gene Aliases: ABLIM3; AW990719; D930036B08Rik; HMFN1661; KIAA0843
UniProt ID: (Human) O94929, (Mouse) Q69ZX8
Entrez Gene ID: (Human) 22885, (Mouse) 319713
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.