Novus Biologicals
Manufacturer Code:NBP237664
Catalog # NBP237664
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1H1 |
Immunogen | Purified recombinant fragment of ABL2 expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Abelson murine leukemia viral (v-abl) oncogene homolog 2 Abelson murine leukemia viral oncogene homolog 2 Abelson-related gene protein Abelson-relatedgene) arg tyrosine kinase EC 2.7.10 EC 2.7.10.2 FLJ22224 FLJ31718 FLJ41441 tyrosine-protein kinase ABL2 Tyrosine-protein kinase ARG v-abl Abelson murine leukemia viral oncogene homolog 2; Abelson murine leukemia viral oncogene homolog 2; Abelson tyrosine-protein kinase 2; Abelson-related gene protein; c-abl oncogene 2, non-receptor tyrosine kinase; Tyrosine-protein kinase ABL2; Tyrosine-protein kinase ARG; v-abl Abelson murine leukemia viral oncogene homolog 2
Gene Aliases: ABL2; ABLL; ARG
UniProt ID: (Human) P42684
Entrez Gene ID: (Human) 27
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.