Invitrogen
Manufacturer Code:PA572165
Catalog # PIPA572165
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 300-329 amino acids from the C-terminal region of mouse Abi3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABI gene family member 3; Nesh; NESH SSH3BP3 ABI gene family member 3 ABI gene family member 3 new molecule including SH3; New molecule including SH3
Gene Aliases: 2210414K06Rik; Abi3; AI987680; NESH
UniProt ID: (Mouse) Q8BYZ1
Entrez Gene ID: (Mouse) 66610
Molecular Function:
G-protein modulator
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.