Novus Biologicals
Manufacturer Code:NBP299345AF750
Catalog # NB084888
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human ABI2. |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Abelson interactor 2; Abelson interactor 2 Abi-2 ABI2B abl binding protein 3 abl interactor 2 Abl-binding protein 3 AblBP3ABI-2 abl-interacting protein 1 (SH3-containing protein) abl-interactor 2 abl-interactor protein 2b AIP-1 arg protein tyrosine kinase-binding protein Arg-binding protein 1 argBP1 ARGBPIA argBPIB SSH3BP2; Abi-2; abl binding protein 3; Abl interactor 2; Abl-binding protein 3; abl-interacting protein 1 (SH3-containing protein); abl-interactor protein 2b; AblBP3; arg protein tyrosine kinase-binding protein; Arg-binding protein 1; ArgBP1
Gene Aliases: ABI-2; ABI2; ABI2B; AblBP3; AIP-1; argBP1; ARGBPIA; argBPIB; SSH3BP2
UniProt ID: (Human) Q9NYB9
Entrez Gene ID: (Human) 10152
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.