Proteintech
Manufacturer Code:148901AP150UL
Catalog # 50-172-9810
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ABI2 Fusion Protein Ag6679 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Abelson interactor 2; Abelson interactor 2 ABI 2 ABI2 ABI2B Abl binding protein 3 abl interactor 2 AblBP3 AIP 1 Arg binding protein 1 ArgBP1 argBPIA argBPIB SSH3BP2; Abi-2; abl binding protein 3; Abl interactor 2; Abl-binding protein 3; abl-interacting protein 1 (SH3-containing protein); abl-interactor protein 2b; AblBP3; arg protein tyrosine kinase-binding protein; Arg-binding protein 1; ArgBP1; thyroid hormone responsive protein
Gene Aliases: 8430425M24Rik; ABI-2; ABI2; ABI2B; AblBP3; AI839867; AIP-1; argBP1; ARGBPIA; argBPIB; C130078H13; SSH3BP2
UniProt ID: (Human) Q9NYB9, (Mouse) P62484
Entrez Gene ID: (Human) 10152, (Mouse) 329165, (Rat) 286928
Molecular Function:
G-protein modulator
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.