Novus Biologicals
Manufacturer Code:NBP257842
Catalog # NBP257842
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SQYGTMTRQISRHNSTTSSTSSGGYRRTPSVTAQFSAQPHVNGGPLYSQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Abelson interactor 1; Abelson interactor 1 ABI-1 abl interactor 1 Abl-binding protein 4 AblBP4 abl-interactor 1 Abl-interactor protein 1 long E3B1 eps8 binding protein Eps8 SH3 domain-binding protein Eps8-binding protein interactor protein AblBP4 nap1 binding protein Nap1-binding protein Nap1BP spectrin SH3 domain binding protein 1 Spectrin SH3 domain-binding protein 1 SSH3BP SSH3BP1; Abi-1; Abl interactor 1; Abl-binding protein 4; abl-interactor 1; Abl-interactor protein 1 long; AblBP4; e3B1; Eps8 SH3 domain-binding protein; Eps8-binding protein; interactor protein AblBP4; nap1 binding protein; Nap1-binding protein; Nap1BP; Spectrin SH3 domain-binding protein 1
Gene Aliases: ABI-1; ABI1; ABLBP4; E3B1; NAP1BP; SSH3BP; SSH3BP1
UniProt ID: (Human) Q8IZP0
Entrez Gene ID: (Human) 10006
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.