Invitrogen
Manufacturer Code:PA536418
Catalog # PIPA536418
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 110-157 of human ABHD9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: abhydrolase domain containing 9; Abhydrolase domain-containing protein 9; EH3; Epoxide hydrolase 3; Epoxide hydrolase 3 Abhydrolase domain-containing protein 9 EPHX3 ABHD9
Gene Aliases: 2310063B19Rik; ABHD9; EH3; EPHX3
UniProt ID: (Human) Q9H6B9, (Mouse) Q3V1F8
Entrez Gene ID: (Human) 79852, (Mouse) 71932, (Rat) 366836
Molecular Function: hydrolase protease serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.