Novus Biologicals
Manufacturer Code:NBP2843790.1ML
Catalog # NB032307
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of mouse ABHD8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: abhydrolase domain containing 8 abhydrolase domain-containing protein 8 EC 3.- FLJ11743 MGC14280 MGC2512; Abhydrolase domain-containing protein 8; Alpha/beta hydrolase domain-containing protein 8; Protein ABHD8
Gene Aliases: ABHD8
UniProt ID: (Human) Q96I13
Entrez Gene ID: (Human) 79575
Molecular Function: hydrolase protease serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.