Invitrogen
Manufacturer Code:PA545761
Catalog # PIPA545761
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human ABHD6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-arachidonoylglycerol hydrolase; abhydrolase domain containing 6; Abhydrolase domain-containing protein 6; lipase protein; Monoacylglycerol lipase ABHD6
Gene Aliases: 0610041D24Rik; AA673485; ABHD6; AV065425
UniProt ID: (Bovine) Q1LZ86, (Human) Q9BV23, (Rat) Q5XI64, (Mouse) Q8R2Y0
Entrez Gene ID: (Bovine) 505283, (Rabbit) 100355792, (Human) 57406, (Dog) 484712, (Rat) 305795, (Guinea pig) 100732636, (Mouse) 66082
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.