Invitrogen
Manufacturer Code:MA538176
Catalog # PIMA538176
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1777 |
Immunogen | A synthesized peptide derived from human ABHD5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 1-acylglycerol-3-phosphate O-acyltransferase ABHD5; 1300003D03Rik 1-acylglycerol-3-phosphate O-acyltransferase ABHD5 2010002J10Rik Abhd5 abhydrolase domain containing 5 abhydrolase domain containing 5 lysophosphatidic acid acyltransferase abhydrolase domain-containing protein 5 abhydrolase domain-containing protein 5 variant 1 abhydrolase domain-containing protein 5 variant 2 aCGI58 CDS CGI 58 CGI58 CGI-58 CGI58 protein IECN2 IECN5 Lipid droplet-binding protein CGI-58 MGC8731 NCIE2 Protein CGI-58 truncated abhydrolase domain-containing protein 5; Abhydrolase domain-containing protein 5; Lipid droplet-binding protein CGI-58; Protein CGI-58; truncated abhydrolase domain-containing protein 5
Gene Aliases: ABHD5; CDS; CGI-58; CGI58; IECN2; NCIE2
UniProt ID: (Human) Q8WTS1, (Rat) Q6QA69
Entrez Gene ID: (Human) 51099, (Rat) 316122
Molecular Function: hydrolase protease serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.