Proteintech
Manufacturer Code:122011AP150UL
Catalog # 50-558-493
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ABHD5 Fusion Protein Ag2840 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-acylglycerol-3-phosphate O-acyltransferase ABHD5; ABHD5 CDS CGI58 IECN2 NCIE2; Abhydrolase domain-containing protein 5; Lipid droplet-binding protein CGI-58; Protein CGI-58; truncated abhydrolase domain-containing protein 5
Gene Aliases: 1300003D03Rik; 2010002J10Rik; ABHD5; CDS; CGI-58; CGI58; IECN2; IECN5; NCIE2
UniProt ID: (Human) Q8WTS1, (Mouse) Q9DBL9
Entrez Gene ID: (Human) 51099, (Mouse) 67469
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.