Invitrogen
Manufacturer Code:PA5116772
Catalog # PIPA5116772
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 223-342 of human ABHD4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (Lyso)-N-acylphosphatidylethanolamine lipase; (Lyso)-N-acylphosphatidylethanolamine lipase 1110035H23Rik ABH4 Abhd4 abhydrolase domain containing 4 abhydrolase domain-containing protein 4 AI429574 Alpha/beta hydrolase domain-containing protein 4 Alpha/beta-hydrolase 4 lyso-N-acylphosphatidylethanolamine lipase protein ABHD4; Abhydrolase domain-containing protein 4; Alpha/beta hydrolase domain-containing protein 4; Alpha/beta-hydrolase 4; lyso-N-acylphosphatidylethanolamine lipase
Gene Aliases: 1110035H23Rik; ABH4; ABHD4; AI429574
UniProt ID: (Human) Q8TB40, (Mouse) Q8VD66
Entrez Gene ID: (Human) 63874, (Rat) 364380, (Mouse) 105501
Molecular Function: hydrolase protease serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.