Invitrogen
Manufacturer Code:PA575764
Catalog # PIPA575764
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human ABHD3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Abhydrolase domain-containing protein 3; Abhydrolase domain-containing protein 3 Lung alpha/beta hydrolase 3 ABHD3 LABH3; alpha/beta hydrolase domain containing protein 3; Lung alpha/beta hydrolase 3; MmLABH3; Phospholipase ABHD3
Gene Aliases: AA675331; ABHD3; LABH3
UniProt ID: (Human) Q8WU67, (Mouse) Q91ZH7
Entrez Gene ID: (Human) 171586, (Rat) 291793, (Mouse) 106861
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.