Invitrogen
Manufacturer Code:PA5103555
Catalog # PIPA5103555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ABHD3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA675331 Abhd3 abhydrolase domain containing 3 abhydrolase domain-containing protein 3 alpha/beta hydrolase domain containing protein 3 Labh3 Lung alpha/beta hydrolase 3 mmLABH3 Phospholipase ABHD3; Abhydrolase domain-containing protein 3; alpha/beta hydrolase domain containing protein 3; Lung alpha/beta hydrolase 3; MmLABH3; Phospholipase ABHD3
Gene Aliases: AA675331; ABHD3; LABH3
UniProt ID: (Human) Q8WU67, (Mouse) Q91ZH7
Entrez Gene ID: (Human) 171586, (Mouse) 106861
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.