Invitrogen
Manufacturer Code:PA5106585
Catalog # PIPA5106585
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ABHD2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-arachidonoylglycerol hydrolase; 2210009N18Rik 2-arachidonoylglycerol hydrolase Abhd2 abhydrolase domain containing 2 abhydrolase domain-containing protein 2 Acetylesterase alpha/beta hydrolase domain containing protein 2 alpha/beta hydrolase fold protein HS1-2 LABH2 Labh-2 Lung alpha/beta hydrolase 2 mmLABH2 Monoacylglycerol lipase ABHD2 PHPS1-2 Progesterone-sensitive lipase Protein PHPS1-2 testicular tissue protein Li 6 Triacylglycerol lipase; Abhydrolase domain-containing protein 2; Acetylesterase; alpha/beta hydrolase domain containing protein 2; alpha/beta hydrolase fold protein; Lung alpha/beta hydrolase 2; MmLABH2; Monoacylglycerol lipase ABHD2; Progesterone-sensitive lipase; Protein PHPS1-2; testicular tissue protein Li 6; Triacylglycerol lipase
Gene Aliases: 2210009N18Rik; ABHD2; HS1-2; Labh-2; LABH2; PHPS1-2
UniProt ID: (Human) P08910, (Mouse) Q9QXM0
Entrez Gene ID: (Human) 11057, (Mouse) 54608
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.