Proteintech
Manufacturer Code:140391AP150UL
Catalog # 50-172-9169
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ABHD2 Fusion Protein Ag5108 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-arachidonoylglycerol hydrolase; ABHD2 HS1 2 LABH2 Lung alpha/beta hydrolase 2 PHPS1 2 Protein PHPS1 2; Abhydrolase domain-containing protein 2; Acetylesterase; alpha/beta hydrolase domain containing protein 2; alpha/beta hydrolase fold protein; Lung alpha/beta hydrolase 2; MmLABH2; Monoacylglycerol lipase ABHD2; Progesterone-sensitive lipase; Protein PHPS1-2; testicular tissue protein Li 6; Triacylglycerol lipase
Gene Aliases: 2210009N18Rik; ABHD2; HS1-2; Labh-2; LABH2; PHPS1-2
UniProt ID: (Human) P08910, (Mouse) Q9QXM0
Entrez Gene ID: (Human) 11057, (Mouse) 54608, (Rat) 293050
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.