Novus Biologicals
Manufacturer Code:NBP2843760.1ML
Catalog # NB032304
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human ABHD18 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: abhydrolase domain-containing protein 18; alpha/beta hydrolase domain-containing protein 18; C4orf29 chromosome 4 open reading frame 29 FLJ21106 hypothetical protein LOC80167 MGC149227 MGC149228; uncharacterized protein C4orf29
Gene Aliases: C4orf29
Entrez Gene ID: (Human) 80167
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.