Novus Biologicals
Manufacturer Code:NBP2825480.1ML
Catalog # NB030477
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human ABHD15 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: abhydrolase domain containing 15 abhydrolase domain-containing protein 15 UNQ6510/PRO21435; Abhydrolase domain-containing protein 15; Alpha/beta hydrolase domain-containing protein 15; Protein ABHD15
Gene Aliases: ABHD15; UNQ6510/PRO21435
UniProt ID: (Human) Q6UXT9
Entrez Gene ID: (Human) 116236
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.