Invitrogen
Manufacturer Code:PA5103554
Catalog # PIPA5103554
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ABHD13 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110065L07Rik ABHD13 abhydrolase domain containing 13 abhydrolase domain-containing protein 13 AI463703 AI788994 alpha/beta hydrolase domain-containing protein 13 bA153I24.2 BEM46L1 C13orf6 Protein ABHD13 RGD1308317 RP11-153I24.2; Abhydrolase domain-containing protein 13; Alpha/beta hydrolase domain-containing protein 13; Protein ABHD13
Gene Aliases: 1110065L07Rik; ABHD13; AI463703; AI788994; bA153I24.2; BEM46L1; C13orf6
UniProt ID: (Human) Q7L211, (Mouse) Q80UX8
Entrez Gene ID: (Human) 84945, (Mouse) 68904
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.