Abnova
Manufacturer Code:PAB17586
Catalog # 89-119-247
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal of human ABHD12B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ABHD12B.
For Research Use Only
Protein Aliases: Abhydrolase domain-containing protein 12B; Alpha/beta hydrolase domain-containing protein 12B; BEM46L3; c14_5314; C14orf29; MGC129926; MGC129927; Protein ABHD12B
Gene Aliases: ABHD12B; BEM46L3; c14_5314; C14orf29
UniProt ID: (Human) Q7Z5M8
Entrez Gene ID: (Human) 145447
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.