Invitrogen
Manufacturer Code:PA518982
Catalog # PIPA518982
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (REFLGKSEPEHQH) corresponding to the C-terminus amino acids of ABHD12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-arachidonoylglycerol hydrolase; 2-arachidonoylglycerol hydrolase ABHD12; ABHD12 abhydrolase domain containing 12 ABHD12A BEM46L2 C20orf22 DKFZP434P106 dJ965G21.2; Abhydrolase domain-containing protein 12; Lysophosphatidylserine lipase ABHD12; Monoacylglycerol lipase ABHD12; Oxidized phosphatidylserine lipase ABHD12
Gene Aliases: 1500011G07Rik; 6330583M11Rik; Abhd12; AI431047; AW547313
UniProt ID: (Mouse) Q99LR1, (Rat) Q6AYT7
Entrez Gene ID: (Mouse) 76192, (Rat) 499913
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.