Invitrogen
Manufacturer Code:PA5101394
Catalog # PIPA5101394
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ABHD12 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1500011G07Rik 2-arachidonoylglycerol hydrolase 2-arachidonoylglycerol hydrolase ABHD12 6330583M11Rik ABHD12 ABHD12A abhydrolase domain containing 12 abhydrolase domain-containing protein 12 AI431047 AW547313 BEM46L2 C20orf22 dJ965G21.2 DKFZP434P106 hABHD12 Lysophosphatidylserine lipase ABHD12 Monoacylglycerol lipase ABHD12 Oxidized phosphatidylserine lipase ABHD12 PHARC; 2-arachidonoylglycerol hydrolase; 2-arachidonoylglycerol hydrolase ABHD12; Abhydrolase domain-containing protein 12; hABHD12; Lysophosphatidylserine lipase ABHD12; Monoacylglycerol lipase ABHD12; Oxidized phosphatidylserine lipase ABHD12
Gene Aliases: 1500011G07Rik; 6330583M11Rik; ABHD12; ABHD12A; AI431047; AW547313; BEM46L2; C20orf22; dJ965G21.2; PHARC
UniProt ID: (Human) A8K450, (Mouse) Q99LR1, (Rat) Q6AYT7
Entrez Gene ID: (Human) 26090, (Mouse) 76192, (Rat) 499913
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.