Novus Biologicals
Manufacturer Code:NBP189308
Catalog # NBP189308
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:ALILESPFTNIREEAKSHPFSVIYRYFPGFDWFFLDPITSSGIKFANDENVKHISCPLLILHAEDDPVVPFQLGRKLYSIAAPARSFRDFKVQFVPFHSDLGYRHKYIYKSPELPRILRP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-arachidonoylglycerol hydrolase; 2-arachidonoylglycerol hydrolase ABHD12; ABHD12A abhydrolase domain containing 12 Abhydrolase domain-containing protein 12 BEM46L2 C20orf22 chromosome 20 open reading frame 22 dJ965G21.2 DKFZP434P106 DKFZp434P1062-arachidonoylglycerol hydrolase EC 3.1.1.23 monoacylglycerol lipase ABHD12 PHARC; Abhydrolase domain-containing protein 12; hABHD12; Lysophosphatidylserine lipase ABHD12; Monoacylglycerol lipase ABHD12; Oxidized phosphatidylserine lipase ABHD12
Gene Aliases: ABHD12; ABHD12A; BEM46L2; C20orf22; dJ965G21.2; PHARC
UniProt ID: (Human) A8K450
Entrez Gene ID: (Human) 26090
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.