Invitrogen
Manufacturer Code:PA5103553
Catalog # PIPA5103553
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ABHD10 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABHD10 abhydrolase domain containing 10 Abhydrolase domain-containing protein 10 abhydrolase domain-containing protein 10 mitochondrial Alpha/beta hydrolase domain-containing protein 10 alpha/beta hydrolase domain-containing protein 10 mitochondrial mycophenolic acid acyl-glucuronide esterase mitochondrial RGD1308084; Abhydrolase domain-containing protein 10; abhydrolase domain-containing protein 10, mitochondrial; Acyl-protein thioesterase ABHD10; Alpha/beta hydrolase domain-containing protein 10; alpha/beta hydrolase domain-containing protein 10, mitochondrial; Mycophenolic acid acyl-glucuronide esterase, mitochondrial; Palmitoyl-protein thioesterase ABHD10, mitochondrial
Gene Aliases: ABHD10; RGD1308084
UniProt ID: (Human) Q9NUJ1, (Rat) Q5I0K5
Entrez Gene ID: (Human) 55347, (Rat) 303953
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.