Novus Biologicals
Manufacturer Code:NBP299171FR
Catalog # NB058622
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human ABHD1. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: abhydrolase domain containing 1 EC 3.1.1 EC 3.1.1.- FLJ36128 LABH1abhydrolase domain-containing protein 1 Lung alpha/beta hydrolase 1; Abhydrolase domain-containing protein 1; Alpha/beta hydrolase domain-containing protein 1; Lung alpha/beta hydrolase 1; Protein ABHD1; testicular tissue protein Li 5
Gene Aliases: ABHD1; LABH1
UniProt ID: (Human) Q96SE0
Entrez Gene ID: (Human) 84696
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.