Proteintech
Manufacturer Code:260911AP150UL
Catalog # 50-173-5203
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ABHD1 Fusion Protein Ag23138 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABHD1 LABH1 Lung alpha/beta hydrolase 1; Abhydrolase domain-containing protein 1; Alpha/beta hydrolase domain-containing protein 1; alpha/beta hydrolase-1; Lung alpha/beta hydrolase 1; MmLABH1; Protein ABHD1; testicular tissue protein Li 5
Gene Aliases: ABHD1; LABH-1; LABH1
UniProt ID: (Human) Q96SE0, (Mouse) Q9QZC8
Entrez Gene ID: (Human) 84696, (Mouse) 57742
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.