Invitrogen
Manufacturer Code:PA597127
Catalog # PIPA597127
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 350-450 of human ABCG5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-binding cassette sub-family G member 5; ATP-binding cassette, sub-family G (WHITE), member 5; ATP-binding cassette, subfamily G, member 5; sterolin 1; Sterolin-1
Gene Aliases: ABCG5; AW112016; cmp; sterolin-1; STSL; trac
UniProt ID: (Human) Q9H222, (Mouse) Q99PE8
Entrez Gene ID: (Human) 64240, (Mouse) 27409
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.