Invitrogen
Manufacturer Code:PA578702
Catalog # PIPA578702
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human ABCG5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-binding cassette sub-family G member 5; ATP-binding cassette sub-family G member 5 Sterolin-1 ABCG5; ATP-binding cassette, sub-family G (WHITE), member 5; ATP-binding cassette, subfamily G, member 5; sterolin 1; Sterolin-1
Gene Aliases: ABCG5; STSL
UniProt ID: (Human) Q9H222
Entrez Gene ID: (Human) 64240
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.